Assay Details
Gene Symbol: OCIAD2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003364714.4 | XP_003364762.1 | 450 | 86 | ||
XM_005608783.3 | XP_005608840.1 | 411 | 86 | |||
XM_005608785.3 | XP_005608842.1 | 364 | 86 | |||
XM_023638114.1 | XP_023493882.1 | 392 | 86 | |||
XM_023638115.1 | XP_023493883.1 | 316 | 86 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
