Assay Details
Gene Symbol: RPL9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001498163.6 | XP_001498213.1 | 267 | 64 | ||
GenBank mRNA | JL626625.1 | - | 2 - 3 | 171 | 64 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
