Assay Details
Gene Symbol: RNF4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001489181.5 | XP_001489231.1 | 792 | 59 | ||
XM_023638492.1 | XP_023494260.1 | 490 | 59 | |||
XM_023638493.1 | XP_023494261.1 | 546 | 59 |

More Information
Set Membership: |
![]() ![]() ![]() |
