Assay Details
Gene Symbol: ADSS2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_023632531.1 | XP_023488299.1 | 879 | 82 | ||
XM_023632532.1 | XP_023488300.1 | 828 | 82 | |||
XM_023632533.1 | XP_023488301.1 | 366 | 82 |

More Information
Set Membership: |
![]() ![]() ![]() |
