Assay Details
Gene Symbol: PLD5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_014738007.2 | XP_014593493.1 | 7600 | 68 | ||
XM_023632621.1 | XP_023488389.1 | 731 | 68 | |||
XM_023632622.1 | XP_023488390.1 | 529 | 68 | |||
XM_023632623.1 | XP_023488391.1 | 202 | 68 |

More Information
Set Membership: |
![]() ![]() ![]() |
