Assay Details
Gene Symbol: ENAH | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005607979.3 | XP_005608036.1 | 1655 | 74 | ||
XM_005607980.3 | XP_005608037.1 | 1653 | 74 | |||
XM_005607981.3 | XP_005608038.1 | 1543 | 74 | |||
XM_014738021.2 | XP_014593507.1 | 1727 | 74 | |||
XM_014738022.2 | XP_014593508.1 | 1715 | 74 | |||
XM_014738026.2 | XP_014593512.1 | 1604 | 74 | |||
XM_023632454.1 | XP_023488222.1 | 1824 | 74 | |||
XM_023632455.1 | XP_023488223.1 | 1921 | 74 | |||
XM_023632456.1 | XP_023488224.1 | 1835 | 74 | |||
XM_023632457.1 | XP_023488225.1 | 1766 | 74 |

More Information
Set Membership: |
![]() ![]() ![]() |
