Assay Details
Gene Symbol: RGS18 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001915707.4 | XP_001915742.1 | 491 | 64 | ||
GenBank mRNA | JL616969.1 | - | 2 - 3 | 501 | 64 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
