Assay Details
Gene Symbol: GLRX2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005608031.3 | XP_005608088.1 | 370 | 93 | ||
XM_023632461.1 | XP_023488229.1 | 449 | 93 | |||
XM_023632462.1 | XP_023488230.1 | 383 | 93 | |||
XM_023632463.1 | XP_023488231.1 | 480 | 93 | |||
XM_023632464.1 | XP_023488232.1 | 400 | 93 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
