Assay Details
Gene Symbol: ADORA1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001496410.6 | XP_001496460.1 | 533 | 100 | ||
XM_005608080.3 | XP_005608137.1 | 840 | 100 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
