Assay Details
Gene Symbol: CHI3L1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005608082.3 | XP_005608139.1 | 433 | 64 | ||
XM_023632449.1 | XP_023488217.1 | 459 | 64 | |||
XM_023632450.1 | XP_023488218.1 | 423 | 64 |

More Information
Set Membership: |
![]() ![]() ![]() |
