Assay Details
Gene Symbol: TFB1M | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001493402.5 | XP_001493452.2 | 601 | 75 | ||
XM_023632971.1 | XP_023488739.1 | 653 | 75 | |||
GenBank mRNA | HQ890135.1 | - | 4 - 5 | 483 | 75 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
