Assay Details
Gene Symbol: MTRF1L | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001501586.4 | XP_001501636.2 | 1078 | 95 | ||
XM_023633084.1 | XP_023488852.1 | 1086 | 95 | |||
GenBank mRNA | JL624917.1 | - | 3 - 4 | 394 | 95 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
