Assay Details
Gene Symbol: ADAT2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_023632867.1 | XP_023488635.1 | 567 | 135 | ||
GenBank mRNA | JL630727.1 | - | 1 - 2 | 186 | 135 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
