Assay Details
Gene Symbol: RALA | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_023638944.1 | XP_023494712.1 | 767 | 100 | ||
XM_023638945.1 | XP_023494713.1 | 552 | 100 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
