Assay Details
Gene Symbol: NNMT | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_014741393.2 | XP_014596879.2 | 734 | 72 | ||
XM_023644850.1 | XP_023500618.1 | 732 | 72 | |||
XM_023644851.1 | XP_023500619.1 | 182 | 72 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
