Assay Details
Gene Symbol: HEPACAM | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_023645077.1 | XP_023500845.1 | 1127 | 65 | ||
XM_023645078.1 | XP_023500846.1 | 1124 | 65 |

More Information
Set Membership: |
![]() ![]() ![]() |
