Assay Details
Gene Symbol: TIRAP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_014741275.2 | XP_014596761.1 | 885 | 80 | ||
XM_023645110.1 | XP_023500878.1 | 2942 | 80 |

More Information
Additional Information:
This assay is in the 5'UTR region of the XM_014741275.2, XM_023645110.1 transcripts.
Set Membership: |
![]() ![]() |
