Assay Details
Gene Symbol: TIRAP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_014741275.2 | XP_014596761.1 | 1172 | 78 | ||
XM_023645110.1 | XP_023500878.1 | 3229 | 78 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: TIRAP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_014741275.2 | XP_014596761.1 | 1172 | 78 | ||
XM_023645110.1 | XP_023500878.1 | 3229 | 78 |