Assay Details
Gene Symbol: ZNF653 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_023645352.1 | XP_023501120.1 | 1221 | 76 | ||
XM_023645354.1 | XP_023501122.1 | 1496 | 76 | |||
XM_023645355.1 | XP_023501123.1 | 1342 | 76 |
