Assay Details
Gene Symbol: ZNF653 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_023645352.1 | XP_023501120.1 | 1539 | 68 | ||
XM_023645353.1 | XP_023501121.1 | 1464 | 68 | |||
XM_023645354.1 | XP_023501122.1 | 1814 | 68 | |||
XM_023645355.1 | XP_023501123.1 | 1660 | 68 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
