Assay Details
Gene Symbol: CDK6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_023639194.1 | XP_023494962.1 | 750 | 64 | ||
XM_023639195.1 | XP_023494963.1 | 779 | 64 | |||
XM_023639196.1 | XP_023494964.1 | 743 | 64 | |||
XM_023639197.1 | XP_023494965.1 | 750 | 64 |

More Information
Set Membership: |
![]() ![]() ![]() |
