Assay Details
Gene Symbol: ASB4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001493628.5 | XP_001493678.1 | 2359 | 73 | ||
XM_023639234.1 | XP_023495002.1 | 1333 | 73 | |||
XM_023639236.1 | XP_023495004.1 | 1196 | 73 | |||
XM_023639237.1 | XP_023495005.1 | 1247 | 73 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
