Assay Details
Gene Symbol: AGR2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001496115.5 | XP_001496165.1 | 249 | 72 | ||
XM_014739196.2 | XP_014594682.1 | 240 | 72 |

More Information
Set Membership: |
![]() ![]() ![]() |
