Assay Details
Gene Symbol: SP4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001497653.5 | XP_001497703.1 | 1837 | 80 | ||
XM_023639346.1 | XP_023495114.1 | 1831 | 80 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: SP4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001497653.5 | XP_001497703.1 | 1837 | 80 | ||
XM_023639346.1 | XP_023495114.1 | 1831 | 80 |