Assay Details
Gene Symbol: SP4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001497653.5 | XP_001497703.1 | 2260 | 78 | ||
XM_023639346.1 | XP_023495114.1 | 2254 | 78 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
