Assay Details
Gene Symbol: GSTK1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001489929.4 | XP_001489979.1 | 507 | 62 | ||
XM_003364870.4 | XP_003364918.1 | 378 | 62 |

More Information
Set Membership: |
![]() ![]() ![]() |
