Assay Details
Gene Symbol: ASPHD2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_023646933.1 | XP_023502701.1 | 1280 | 106 | ||
XM_023646934.1 | XP_023502702.1 | 2133 | 106 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
