Assay Details
Gene Symbol: GLTP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001496768.5 | XP_001496818.3 | 391 | 81 | ||
XM_005612486.3 | XP_005612543.2 | 325 | 81 |

More Information
Set Membership: |
![]() ![]() ![]() |
