Assay Details
Gene Symbol: PLBD2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_023647111.1 | XP_023502879.1 | 681 | 62 | ||
XM_023647112.1 | XP_023502880.1 | 682 | 62 |

More Information
Set Membership: |
![]() ![]() ![]() |
