Assay Details
Gene Symbol: CIDEA | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001489777.5 | XP_001489827.2 | 417 | 84 | ||
XM_005612753.3 | XP_005612810.1 | 417 | 84 | |||
XM_014727780.2 | XP_014583266.1 | 598 | 84 |
