Assay Details
Gene Symbol: LOC100058979 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001490037.6 | XP_001490087.1 | 723 | 106 | ||
XM_005612762.3 | XP_005612819.1 | 878 | 106 |

More Information
Set Membership: |
![]() ![]() |
