Assay Details
Gene Symbol: SS18 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_023647591.1 | XP_023503359.1 | 318 | 104 | ||
XM_023647592.1 | XP_023503360.1 | 184 | 104 | |||
XM_023647593.1 | XP_023503361.1 | 317 | 104 | |||
XM_023647594.1 | XP_023503362.1 | 318 | 104 |

More Information
Set Membership: |
![]() ![]() ![]() |
