Assay Details
Gene Symbol: GAREM1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001496712.6 | XP_001496762.4 | 375 | 75 | ||
XM_005612821.3 | XP_005612878.2 | 375 | 75 | |||
XM_023647616.1 | XP_023503384.1 | 385 | 75 |
