Assay Details
Gene Symbol: INO80C | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001496752.6 | XP_001496802.1 | 324 | 107 | ||
XM_005612842.3 | XP_005612899.1 | 345 | 107 | |||
XM_005612843.3 | XP_005612900.1 | 337 | 107 | |||
XM_023647661.1 | XP_023503429.1 | 346 | 107 | |||
GenBank mRNA | JL639404.1 | - | 1 - 2 | 94 | 107 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
