Assay Details
Gene Symbol: KIAA1328 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001497549.5 | XP_001497599.4 | 814 | 82 | ||
XM_005612850.3 | XP_005612907.2 | 814 | 82 | |||
XM_005612851.3 | XP_005612908.2 | 601 | 82 | |||
XM_014727689.2 | XP_014583175.2 | 1517 | 82 | |||
XM_023648260.1 | XP_023504028.1 | 580 | 82 | |||
XM_023648261.1 | XP_023504029.1 | 588 | 82 | |||
XM_023648262.1 | XP_023504030.1 | 814 | 82 |

More Information
Set Membership: |
![]() ![]() ![]() |
