Assay Details
Gene Symbol: MAPK4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_023647815.1 | XP_023503583.1 | 2093 | 65 | ||
XM_023647816.1 | XP_023503584.1 | 2298 | 65 | |||
XM_023647817.1 | XP_023503585.1 | 1931 | 65 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
