Assay Details
Gene Symbol: BCL2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001490436.4 | XP_001490486.1 | 820 | 102 | ||
XM_023647939.1 | XP_023503707.1 | 843 | 102 |

More Information
Additional Information:
This assay is in the 5'UTR region of the XM_001490436.4, XM_023647939.1 transcripts.
Set Membership: |
![]() ![]() ![]() |
