Assay Details
Gene Symbol: SERPINB8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001492201.4 | XP_001492251.1 | 554 | 64 | ||
XM_014727840.2 | XP_014583326.1 | 452 | 64 | |||
XM_023647953.1 | XP_023503721.1 | 735 | 64 | |||
XM_023647954.1 | XP_023503722.1 | 1603 | 64 |

More Information
Set Membership: |
![]() ![]() ![]() |
