Assay Details
Gene Symbol: TERF1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001492421.6 | XP_001492471.1 | 1174 | 92 | ||
XM_001492463.6 | XP_001492513.1 | 1115 | 92 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
