Assay Details
Gene Symbol: DNAJC5B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005613089.3 | XP_005613146.1 | 908 | 66 | ||
XM_014728010.2 | XP_014583496.1 | 775 | 66 | |||
XM_023648526.1 | XP_023504294.1 | 604 | 66 | |||
XM_023648527.1 | XP_023504295.1 | 1010 | 66 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
