Assay Details
Gene Symbol: LYPLA1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005613164.3 | XP_005613221.3 | 233 | 96 | ||
XM_023648610.1 | XP_023504378.1 | 512 | 96 | |||
XM_023648611.1 | XP_023504379.1 | 513 | 96 | |||
XM_023648612.1 | XP_023504380.1 | 1353 | 96 | |||
XM_023648613.1 | XP_023504381.1 | 485 | 96 |

More Information
Set Membership: |
![]() ![]() ![]() |
