Assay Details
Gene Symbol: DECR1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001488668.4 | XP_001488718.2 | 719 | 74 | ||
XM_005613191.3 | XP_005613248.1 | 787 | 74 | |||
XM_023648666.1 | XP_023504434.1 | 680 | 74 |

More Information
Set Membership: |
![]() ![]() ![]() |
