Assay Details
Gene Symbol: CPQ | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001490623.5 | XP_001490673.1 | 1295 | 64 | ||
XM_023648737.1 | XP_023504505.1 | 847 | 64 | |||
XM_023648738.1 | XP_023504506.1 | 902 | 64 |

More Information
Set Membership: |
![]() ![]() ![]() |
