Assay Details
Gene Symbol: OSR2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005613226.3 | XP_005613283.3 | 1321 | 128 | ||
XM_023648756.1 | XP_023504524.1 | 1254 | 128 | |||
XM_023648757.1 | XP_023504525.1 | 1393 | 128 | |||
XM_023648758.1 | XP_023504526.1 | 1546 | 128 | |||
XM_023648760.1 | XP_023504528.1 | 1068 | 128 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
