Assay Details
Gene Symbol: LRP12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001494687.5 | XP_001494737.1 | 310 | 89 | ||
XM_003365614.4 | XP_003365662.1 | 250 | 89 |

More Information
Set Membership: |
![]() ![]() ![]() |
