Assay Details
Gene Symbol: MYC | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001497991.5 | XP_001498041.2 | 1292 | 101 | ||
XM_014728081.2 | XP_014583567.1 | 1300 | 101 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
