Assay Details
Gene Symbol: GLI4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005613336.3 | XP_005613393.1 | 3363 | 101 | ||
XM_023649110.1 | XP_023504878.1 | 673 | 101 | |||
XM_023649111.1 | XP_023504879.1 | 585 | 101 | |||
XM_023649112.1 | XP_023504880.1 | 679 | 101 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
