Assay Details
Gene Symbol: SLC52A2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001504999.5 | XP_001505049.1 | 1314 | 84 | ||
XM_014728125.2 | XP_014583611.1 | 1244 | 84 | |||
XM_023649298.1 | XP_023505066.1 | 1213 | 84 | |||
XM_023649299.1 | XP_023505067.1 | 1232 | 84 | |||
XM_023649300.1 | XP_023505068.1 | 1255 | 84 | |||
XM_023649301.1 | XP_023505069.1 | 1310 | 84 | |||
XM_023649302.1 | XP_023505070.1 | 1159 | 84 |

More Information
Set Membership: |
![]() ![]() ![]() |
