Assay Details
Gene Symbol: MFSD3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001495501.4 | XP_001495551.3 | 1448 | 89 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: MFSD3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001495501.4 | XP_001495551.3 | 1448 | 89 |