Assay Details
Gene Symbol: DYNC2I1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_023640125.1 | XP_023495893.1 | 2341 | 62 | ||
XM_023640126.1 | XP_023495894.1 | 2241 | 62 | |||
XM_023640127.1 | XP_023495895.1 | 3836 | 62 | |||
XM_023640128.1 | XP_023495896.1 | 1816 | 62 | |||
XM_023640129.1 | XP_023495897.1 | 938 | 62 | |||
XM_023640130.1 | XP_023495898.1 | 2497 | 62 | |||
XM_023640131.1 | XP_023495899.1 | 1029 | 62 |

More Information
Set Membership: |
![]() ![]() ![]() |
